MilliporeSigma
  • n-3 Fatty acids from fish or fish-oil supplements, but not alpha-linolenic acid, benefit cardiovascular disease outcomes in primary- and secondary-prevention studies: a systematic review.
n-3 Fatty acids from fish or fish-oil supplements, but not alpha-linolenic acid, benefit cardiovascular disease outcomes in primary- and secondary-prevention studies: a systematic review.
The American journal of clinical nutrition (2006-07-11)
Chenchen Wang, William S Harris, Mei Chung, Alice H Lichtenstein, Ethan M Balk, Bruce Kupelnick, Harmon S Jordan, Joseph Lau
ABSTRACT
Studies on the relation between dietary n-3 fatty acids (FAs) and cardiovascular disease vary in quality, and the results are inconsistent. A systematic review of the literature on the effects of n-3 FAs (consumed as fish or fish oils rich in eicosapentaenoic acid and docosahexaenoic acid or as alpha-linolenic acid) on cardiovascular disease outcomes and adverse events was conducted. Studies from MEDLINE and other sources that were of > or =1 y in duration and that reported estimates of fish or n-3 FA intakes and cardiovascular disease outcomes were included. Secondary prevention was addressed in 14 randomized controlled trials (RCTs) of fish-oil supplements or of diets high in n-3 FAs and in 1 prospective cohort study. Most trials reported that fish oil significantly reduced all-cause mortality, myocardial infarction, cardiac and sudden death, or stroke. Primary prevention of cardiovascular disease was reported in 1 RCT, in 25 prospective cohort studies, and in 7 case-control studies. No significant effect on overall deaths was reported in 3 RCTs that evaluated the effects of fish oil in patients with implantable cardioverter defibrillators. Most cohort studies reported that fish consumption was associated with lower rates of all-cause mortality and adverse cardiac outcomes. The effects on stroke were inconsistent. Evidence suggests that increased consumption of n-3 FAs from fish or fish-oil supplements, but not of alpha-linolenic acid, reduces the rates of all-cause mortality, cardiac and sudden death, and possibly stroke. The evidence for the benefits of fish oil is stronger in secondary- than in primary-prevention settings. Adverse effects appear to be minor.
Product Description
Supelco
Linolenic acid, analytical standard
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Sigma-Aldrich
Linolenic acid, ≥99%