MilliporeSigma
  • Bicalutamide 150mg: a review of its use in the treatment of locally advanced prostate cancer.
Bicalutamide 150mg: a review of its use in the treatment of locally advanced prostate cancer.
Drugs (2006-05-19)
Keri Wellington, Susan J Keam
ABSTRACT
Bicalutamide (Casodex) is a competitive androgen receptor antagonist that inactivates androgen-regulated prostate cell growth and function, leading to cell apoptosis and inhibition of prostate cancer growth. It is administered orally as a once-daily dose. In the EU and a number of other countries, bicalutamide 150 mg/day is approved in men with locally advanced nonmetastatic prostate cancer as immediate therapy either as an adjuvant to active treatment or as monotherapy as an alternative to surgical or medical castration. Combined analysis of the three trials that comprise the bicalutamide Early Prostate Cancer (EPC) programme showed that bicalutamide administered in conjunction with standard care in men with locally advanced prostate cancer offers disease-free survival benefits over standard care alone and is generally well tolerated. Overall survival was improved to a greater extent in the subgroup of patients who received bicalutamide plus radiation therapy compared with radiation therapy alone. Men with localised prostate cancer do not benefit from the addition of bicalutamide to standard care. Combined analysis of two other studies in men with locally advanced prostate cancer show that bicalutamide monotherapy offers better tolerability and higher health-related quality-of-life (HR-QOL) scores for sexual interest and physical capacity compared with surgical or medical castration, while achieving disease-free and overall survival durations that were not significantly different. Thus, when treatment options are being evaluated, bicalutamide as adjuvant therapy or monotherapy should be considered as an alternative to other available hormonal therapies in men with locally advanced prostate cancer, especially in those who wish to maintain an active lifestyle.
Sigma-Aldrich
Bicalutamide (CDX), ≥98% (HPLC), powder
Bicalutamide for system suitability, European Pharmacopoeia (EP) Reference Standard
Bicalutamide, European Pharmacopoeia (EP) Reference Standard