MilliporeSigma
Meloxicam.
Expert opinion on pharmacotherapy (2002-10-22)
Roy Fleischmann, Imran Iqbal, Gleb Slobodin
ABSTRACT
Meloxicam (Mobic trade mark, Boehringer Ingelheim) is a relatively new oral non-steroidal anti-inflammatory drug (NSAID) approved for the treatment of osteoarthritis in the US. It has also been evaluated for the treatment of rheumatoid arthritis, ankylosing spondylitis and acute 'rheumatic' pain. Meloxicam has been shown to be COX-2 preferential, particularly at its lowest therapeutic dose, and is anti-inflammatory by inhibiting prostanoid synthesis in inflammatory cells. Since it is COX-2 preferential, it would be expected to have less gastrointestinal toxicity than non-selective NSAIDs. In clinical trials of meloxicam in osteoarthritis, it was found to be as effective as piroxicam, diclofenac and naproxen with less clinical gastrointestinal symptoms and less perforations, obstructions and bleeds by meta-analysis. Adverse events, including peripheral oedema and hypertension, occurred at a similar rate as with traditional NSAIDs.
Product Description
USP
Meloxicam, United States Pharmacopeia (USP) Reference Standard
Meloxicam, European Pharmacopoeia (EP) Reference Standard