MilliporeSigma
  • Asymmetric Au-catalyzed cycloisomerization of 1,6-enynes: An entry to bicyclo[4.1.0]heptene.
Asymmetric Au-catalyzed cycloisomerization of 1,6-enynes: An entry to bicyclo[4.1.0]heptene.
Beilstein journal of organic chemistry (2011-09-15)
Alexandre Pradal, Chung-Meng Chao, Patrick Y Toullec, Véronique Michelet
ABSTRACT
A comprehensive study on the asymmetric gold-catalyzed cycloisomerization reaction of heteroatom tethered 1,6-enynes is described. The cycloisomerization reactions were conducted in the presence of the chiral cationic Au(I) catalyst consisting of (R)-4-MeO-3,5-(t-Bu)(2)-MeOBIPHEP-(AuCl)(2) complex and silver salts (AgOTf or AgNTf(2)) in toluene under mild conditions to afford functionalized bicyclo[4.1.0]heptene derivatives. The reaction conditions were found to be highly substrate-dependent, the best results being obtained in the case of oxygen-tethered enynes. The formation of bicyclic derivatives, including cyclopropyl pentasubstituted ones, was reported in moderate to good yields and in enantiomeric excesses up to 99%.
Sigma-Aldrich
3-Bromoiodobenzene, 98%