MilliporeSigma
  • Pancreatic islet enhancer clusters enriched in type 2 diabetes risk-associated variants.
Pancreatic islet enhancer clusters enriched in type 2 diabetes risk-associated variants.
Nature genetics (2014-01-15)
Lorenzo Pasquali, Kyle J Gaulton, Santiago A Rodríguez-Seguí, Loris Mularoni, Irene Miguel-Escalada, İldem Akerman, Juan J Tena, Ignasi Morán, Carlos Gómez-Marín, Martijn van de Bunt, Joan Ponsa-Cobas, Natalia Castro, Takao Nammo, Inês Cebola, Javier García-Hurtado, Miguel Angel Maestro, François Pattou, Lorenzo Piemonti, Thierry Berney, Anna L Gloyn, Philippe Ravassard, José Luis Gómez Skarmeta, Ferenc Müller, Mark I McCarthy, Jorge Ferrer
ABSTRACT
Type 2 diabetes affects over 300 million people, causing severe complications and premature death, yet the underlying molecular mechanisms are largely unknown. Pancreatic islet dysfunction is central in type 2 diabetes pathogenesis, and understanding islet genome regulation could therefore provide valuable mechanistic insights. We have now mapped and examined the function of human islet cis-regulatory networks. We identify genomic sequences that are targeted by islet transcription factors to drive islet-specific gene activity and show that most such sequences reside in clusters of enhancers that form physical three-dimensional chromatin domains. We find that sequence variants associated with type 2 diabetes and fasting glycemia are enriched in these clustered islet enhancers and identify trait-associated variants that disrupt DNA binding and islet enhancer activity. Our studies illustrate how islet transcription factors interact functionally with the epigenome and provide systematic evidence that the dysregulation of islet enhancers is relevant to the mechanisms underlying type 2 diabetes.
Sigma-Aldrich
Anti-CTCF Antibody, serum, Upstate®