MilliporeSigma
Synthesis of diethynyltriptycene-linked dipyridyl ligands.
Organic letters (2005-10-08)
Jeremy J Kodanko, Anna J Morys, Stephen J Lippard
ABSTRACT
[reaction: see text] An efficient route to a new family of dinucleating ligands has been developed. A convergent strategy to these ligands involved dual Sonogashira cross-coupling of 2,3-diethynyltriptycene with a variety of functionally diverse 5-bromopyridines. The resultant ligands were accessed in four steps and 40-50% overall yield from 1,2,4,5-tetrabromobenzene. Synthesis of an imidazole and a quinoline derivative by this method is also described.
Sigma-Aldrich
1,2,4,5-Tetrabromobenzene, 97%