MilliporeSigma
  • (3-substituted benzyl)thiazolidine-2,4-diones as structurally new antihyperglycemic agents.
(3-substituted benzyl)thiazolidine-2,4-diones as structurally new antihyperglycemic agents.
Bioorganic & medicinal chemistry letters (1999-03-31)
M Nomura, S Kinoshita, H Satoh, T Maeda, K Murakami, M Tsunoda, H Miyachi, K Awano
ABSTRACT
A series of 3-[(2,4-dioxothiazolidin-5-yl)methyl]benzamide derivatives was prepared as part of a search for antidiabetic agents. A structure-activity relationship study of these compounds led to the identification of 5-[(2,4-dioxothiazolidin-5-yl)methyl]-2-methoxy-N-[[4-(trifluorome thyl)-phenyl]methyl]benzamide (KRP-297) as a candidate drug for the treatment of diabetes mellitus.
Sigma-Aldrich
4-(Trifluoromethyl)benzylamine, 97%