MilliporeSigma
  • A study of common interferences with the forensic luminol test for blood.
A study of common interferences with the forensic luminol test for blood.
Luminescence : the journal of biological and chemical luminescence (2001-08-21)
T I Quickenden, J I Creamer
ABSTRACT
A wide range of domestic and industrial substances that might be mistaken for haemoglobin in the forensic luminol test for blood were examined. The substances studied were in the categories of vegetable or fruit pulps and juices; domestic and commercial oils; cleaning agents; an insecticide; and various glues, paints and varnishes. A significant number of substances in each category gave luminescence intensities that were comparable with the intensities of undiluted haemoglobin, when sprayed with the standard forensic solution containing aqueous alkaline luminol and sodium perborate. In these cases the substance could be easily mistaken for blood when the luminol test is used, but in the remaining cases the luminescence intensity was so weak that it is unlikely that a false-positive test would be obtained. In a few cases the brightly emitting substance could be distinguished from blood by a small but detectable shift of the peak emission wavelength. The results indicated that particular care should be taken to avoid interferences when a crime scene is contaminated with parsnip, turnip or horseradish, and when surfaces coated with enamel paint are involved. To a lesser extent, some care should be taken when surfaces covered with terracotta or ceramic tiles, polyurethane varnishes or jute and sisal matting are involved.
Sigma-Aldrich
Luminol, 97%
Pricing and availability is not currently available.