MilliporeSigma
The design and synthesis of inhibitors of pantothenate synthetase.
Organic & biomolecular chemistry (2006-09-23)
Kellie L Tuck, S Adrian Saldanha, Louise M Birch, Alison G Smith, Chris Abell
ABSTRACT
Pantothenate synthetase catalyses the ATP-dependent condensation of D-pantoate and beta-alanine to form pantothenate. Ten analogues of the reaction intermediate pantoyl adenylate, in which the phosphodiester is replaced by either an ester or sulfamoyl group, were designed as potential inhibitors of the enzyme. The esters were all modest competitive inhibitors, the sulfamoyls were more potent, consistent with their closer structural similarity to the pantoyl adenylate intermediate.
Product Description
Supelco
Diaion® HP-20SS, bucket of 1000 g
Pricing and availability is not currently available.
Supelco
Diaion® HP-20SS, jar of 100 g
Pricing and availability is not currently available.