MilliporeSigma
  • Combinatorial chemistry: from peptides and peptidomimetics to small organic and heterocyclic compounds.
Combinatorial chemistry: from peptides and peptidomimetics to small organic and heterocyclic compounds.
Bioorganic & medicinal chemistry letters (1999-01-05)
A Nefzi, C Dooley, J M Ostresh, R A Houghten
ABSTRACT
Modified dipeptides have been used successfully for the generation of a variety of small organic and heterocyclic combinatorial libraries, including linear urea, polyamine, hydantoin, thiohydantoin, cyclic urea, cyclic thiourea and bicyclic guanidine. The synthesis and screening results for a number of these libraries are described. The solid phase synthesis of heterocyclic compounds such as diazepine and thiomorpholinone are also described.