MilliporeSigma
  • Synthesis and evaluation of urea and thiourea derivatives of oxazolidinones as antibacterial agents.
Synthesis and evaluation of urea and thiourea derivatives of oxazolidinones as antibacterial agents.
Chemical & pharmaceutical bulletin (2007-02-03)
Sunil Kumar Reddy Aaramadaka, Mrinal Kanthi Guha, Ganesh Prabhu, Suvarna Ganesh Kini, Magesh Vijayan
ABSTRACT
Urea and thiourea derivatives of oxazolidinones were synthesized and their inhibitory activity (MIC) was determined on the bacterial strains which includes clinical isolates and quality control organisms. The structure activity relationships were studied and a 3D-QSAR model was built using Genetic Function Approximation. Interestingly found that electron withdrawing groups at the ortho position of the phenyl ring enhances the activity.
Sigma-Aldrich
3-Bromophenyl isothiocyanate, 97%