MilliporeSigma
  • Complexome profiling identifies TMEM126B as a component of the mitochondrial complex I assembly complex.
Complexome profiling identifies TMEM126B as a component of the mitochondrial complex I assembly complex.
Cell metabolism (2012-09-18)
Heinrich Heide, Lea Bleier, Mirco Steger, Jörg Ackermann, Stefan Dröse, Bettina Schwamb, Martin Zörnig, Andreas S Reichert, Ina Koch, Ilka Wittig, Ulrich Brandt
ABSTRACT
Macromolecular complexes are essential players in numerous biological processes. They are often large, dynamic, and rather labile; approaches to study them are scarce. Covering masses up to ∼30 MDa, we separated the native complexome of rat heart mitochondria by blue-native and large-pore blue-native gel electrophoresis to analyze its constituents by mass spectrometry. Similarities in migration patterns allowed hierarchical clustering into interaction profiles representing a comprehensive analysis of soluble and membrane-bound complexes of an entire organelle. The power of this bottom-up approach was validated with well-characterized mitochondrial multiprotein complexes. TMEM126B was found to comigrate with known assembly factors of mitochondrial complex I, namely CIA30, Ecsit, and Acad9. We propose terming this complex mitochondrial complex I assembly (MCIA) complex. Furthermore, we demonstrate that TMEM126B is required for assembly of complex I. In summary, complexome profiling is a powerful and unbiased technique allowing the identification of previously overlooked components of large multiprotein complexes.
Sigma-Aldrich
Anti-ACAD9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution