MilliporeSigma
  • Preparation of highly functionalized alkylzinc halides from alkyl bromides using Mg, ZnCl2 and LiCl.
Preparation of highly functionalized alkylzinc halides from alkyl bromides using Mg, ZnCl2 and LiCl.
Chemical communications (Cambridge, England) (2010-04-10)
Tobias D Blümke, Fabian M Piller, Paul Knochel
ABSTRACT
Alkyl bromides bearing various functional groups are smoothly converted to functionalized alkylzinc reagents by the direct insertion of magnesium in the presence of LiCl and ZnCl(2).
Sigma-Aldrich
Copper(I) cyanide di(lithium chloride) complex solution, in anhydrous tetrahydrofuran