MilliporeSigma
  • Effects of carcinogen and/or mutagen on normal and gonatotropin-primed ovaries of mice.
Effects of carcinogen and/or mutagen on normal and gonatotropin-primed ovaries of mice.
International journal of cancer (1981-07-15)
A R Rao
ABSTRACT
The influence of pregnant mare serum gonadotropin (PSGM) on the induction of ovarian tumors by carcinogen(DMBA) and/or mutagens (EMS and BMS) has been studied in Swiss albino mice. Priming of the ovaries of 6-week-old mice with PMSG (50 IU/mouse) 24 h before intragastric intubation of DMBA (5 mg/mouse) resulted in a significant increase in ovarian tumors when harvested 6 months after the treatment. A similar experiment done on 8-week-old mice yielded confirmatory results. Mutagens (EMS and BMS) failed, at given dose levels (200 mg/kg and 300 mg/kg body weight), to induce tumors in normal as well as PMSG-primed ovaries. Pretreatment of animals with PMSG-enhanced DMBA-induced oocyte depletion and also increased the DMBA-3H activity in the ovaries. The augmentation of DMBA-induced tumors in PMSG-primed ovaries could be due to increased uptake of DMBA and increased depletion of oocytes. It is also possible that PMSG-primed ovaries provided more proliferating granulosa cells (and macromolecules) for the interaction of DMBA (or its metabolites) and thereby increased the chances of cell transformation in the ovaries.
Product Description
Butyl methanesulfonate, European Pharmacopoeia (EP) Reference Standard