MilliporeSigma
  • [The behavior of blood clotting and its inhibitors under long term treatment with 5,6-benzo-alpha-pyrone (coumarin). Double blind study].
[The behavior of blood clotting and its inhibitors under long term treatment with 5,6-benzo-alpha-pyrone (coumarin). Double blind study].
Arzneimittel-Forschung (1985-01-01)
H Köstering, B Bandura, H A Merten, J U Wieding
ABSTRACT
The analyses on hemostaseological variables are recorded in connection with blood tests in the course of a double-blind study in 41 patients suffering from chronic venous insufficiency of higher degrees of severity. They were treated in addition to compression measures with two active ingredients, a coumarin (5,6-benzo-alpha-pyrone)/troxerutin combination (Venalot Depot) (n = 20) or benzarone (n = 21) receiving 3 X 2 dragees daily for 6 weeks. Good clinical efficacy and the improvement of symptoms were observed together with almost no side-effects. The coagulation analysis showed no influence of the active principles on the global coagulation or the clotting factors and the inhibitors and factors of the fibrinolysis. In particular there was no phenprocoumon-like effect on the blood clotting system.
Product Description
Benzarone, European Pharmacopoeia (EP) Reference Standard