MilliporeSigma
  • Semisynthetic pyrrolizidine alkaloid N-oxide antitumor agents. Esters of heliotridine.
Semisynthetic pyrrolizidine alkaloid N-oxide antitumor agents. Esters of heliotridine.
Journal of medicinal chemistry (1988-08-01)
L H Zalkow, J A Glinski, L T Gelbaum, D Moore, D Melder, G Powis
ABSTRACT
The C-9 and C-7 monoesters and C-7, C-9 diesters of heliotridine with (S)-(+) and (R)-(-)-2-hydroxy-2-phenylbutyric acid were prepared, converted into their N-oxides, and compared with the corresponding C-9 monoesters of retronecine in the in vivo P388 lymphocytic leukemia screen. Relative in vitro cytotoxicities of some of the free bases and their corresponding N-oxides were also measured against the A204 rhabdomyosarcoma cell line by using the soft agar colony forming assay. Stereochemistry at C-7 of the necine and at C-2' of the necine acid appears to have a significant effect on the antitumor activity in this system. In the heliotridine series, the configuration of the necic acid has a pronounced effect on the site selectivity (C-7 vs C-9) in esterification with carbodiimidazole. An explanation for this site selectivity is offered.