MilliporeSigma
  • The urinary excretion of frusemide and its metabolites by kidney transplant patients.
The urinary excretion of frusemide and its metabolites by kidney transplant patients.
European journal of clinical pharmacology (1987-01-01)
H Nakahama, Y Miwa, A Yamaji, Y Orita, Y Fukuhara, M Yanase, T Kamada, T Sonoda, M Ishibasi, Y Ichikawa
ABSTRACT
Urine from 5 renal transplant recipients treated with frusemide was analyzed for unchanged frusemide (F), glucuronidated frusemide (G) and 4-chloro-5-sulfamoylanthranilic acid (CSA) by HPLC. In 3 recipients, whose renal function recovered steadily and whose hepatic function was normal throughout, the ratio of frusemide to its metabolites, F/(F + G + CSA), increased steadily in conjunction with the recovery of renal function. In one patient, who received frusemide 200-400 mg/day i.v., the urinary CSA concentration was 64-102 micrograms X ml-1. In 2 patients who experienced shock and/or hepatic dysfunction after transplantation, the F/(F + G + CSA) ratio fluctuated.
Product Description
USP
Furosemide Related Compound B, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.