MilliporeSigma
  • Androgen therapy in aplastic anaemia: a comparative study of high and low-doses and of 4 different androgens. French Cooperative Group for the Study of Aplastic and Refractory Anemias.
Androgen therapy in aplastic anaemia: a comparative study of high and low-doses and of 4 different androgens. French Cooperative Group for the Study of Aplastic and Refractory Anemias.
Scandinavian journal of haematology (1986-04-01)
ABSTRACT
A prospective randomized study of androgen therapy in aplastic anaemia (AA) was performed: 2 androgens (fluoxymesterone and norethandrolone) at high (1 mg/kg/d) and low (0.2 mg/kg/d) dose were studied on 110 patients; and 4 androgens given at high doses were objectively compared in 125 other cases. When patients are matched for AA severity there is an obvious efficiency of the high-dose androgens in the survival of the less severe cases of aplastic anaemia and in their haematological improvement. The data further show that, among the 4 androgens tested, fluoxymesterone is the most efficient and stanozolol the least. We conclude that androgen therapy is truly efficient in moderately severe aplastic anaemia and that fluoxymesterone (1 mg/kg/d) for 18 months should be chosen as reference drug to any new androgen therapy assay.