MilliporeSigma
  • A randomised controlled trial to compare intravenous iron sucrose and oral iron in treatment of iron deficiency anemia in pregnancy.
A randomised controlled trial to compare intravenous iron sucrose and oral iron in treatment of iron deficiency anemia in pregnancy.
Indian journal of hematology & blood transfusion : an official journal of Indian Society of Hematology and Blood Transfusion (2014-05-20)
Avantika Gupta, Usha Manaktala, Asmita Muthal Rathore
ABSTRACT
The aim of this study was to compare the efficacy and safety of intravenous iron sucrose with oral iron therapy in pregnant patients with anemia. The primary outcome of the study was increase in haemoglobin on day 7, 14 & 28 and rise of serum ferritin over 28 days. The study population consisted of 100 patients with singleton pregnancy between 24 and 34 weeks, hemoglobin levels between 7.0-9.0 gm/dL and serum ferritin levels less than 15 ng/mL. The participants in the oral group were given daily 180 mg elemental iron in three divided oral doses for 4 weeks. Total calculated dose of iron sucrose with a target hemoglobin of 11 gm %, was given in 200 mg dose on alternate days. Mean haemoglobin rise was 0.58 gm/dL in the IV group as compared to 0.23 gm/dL in the oral group on day 14 and 1.9 gm/dL in the IV group & 1.3 gm/dL in the oral group on day 28, (p <0.05). In the IV group, 76% of the subjects achieved haemoglobin levels of ≥11 gm% at the time of delivery, as compared to only 54% of the subjects in the oral group who achieved these levels. Serum ferritin value was significantly higher in the IV group, 37.45 ± 5.73 ng/mL as compared to 13.96 ± 1.88 ng/mL in the oral group at 4th week (p <0.001). There was no major side effect in the IV group. 36% subjects in the oral group developed gastrointestinal side effects & 10% of the subjects were non compliant. The rate of hemoglobin rise is faster with intravenous iron sucrose therapy as compared to oral iron therapy which can be beneficial in pregnant women presenting with anemia at a later period of gestation. Intravenous iron sucrose is very well tolerated during pregnancy.
Product Description
Supelco
3,3′,5-Triiodo-L-thyronine-13C6 solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®