MilliporeSigma
  • Impaired D-serine-mediated cotransmission mediates cognitive dysfunction in epilepsy.
Impaired D-serine-mediated cotransmission mediates cognitive dysfunction in epilepsy.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-08-09)
Katharina Klatte, Timo Kirschstein, David Otte, Leonie Pothmann, Lorenz Müller, Tursonjan Tokay, Maria Kober, Mischa Uebachs, Andreas Zimmer, Heinz Beck
ABSTRACT
The modulation of synaptic plasticity by NMDA receptor (NMDAR)-mediated processes is essential for many forms of learning and memory. Activation of NMDARs by glutamate requires the binding of a coagonist to a regulatory site of the receptor. In many forebrain regions, this coagonist is d-serine. Here, we show that experimental epilepsy in rats is associated with a reduction in the CNS levels of d-serine, which leads to a desaturation of the coagonist binding site of synaptic and extrasynaptic NMDARs. In addition, the subunit composition of synaptic NMDARs changes in chronic epilepsy. The desaturation of NMDARs causes a deficit in hippocampal long-term potentiation, which can be rescued with exogenously supplied d-serine. Importantly, exogenous d-serine improves spatial learning in epileptic animals. These results strongly suggest that d-serine deficiency is important in the amnestic symptoms of temporal lobe epilepsy. Our results point to a possible clinical utility of d-serine to alleviate these disease manifestations.