MilliporeSigma
  • Asymmetric construction of all-carbon quaternary stereocenters by chiral-auxiliary-mediated Claisen rearrangement and total synthesis of (+)-bakuchiol.
Asymmetric construction of all-carbon quaternary stereocenters by chiral-auxiliary-mediated Claisen rearrangement and total synthesis of (+)-bakuchiol.
Molecules (Basel, Switzerland) (2012-11-10)
Ken-ichi Takao, Shu Sakamoto, Marianne Ayaka Touati, Yusuke Kusakawa, Kin-ichi Tadano
ABSTRACT
An asymmetric Claisen rearrangement using Oppolzer’s camphorsultam was developed. Under thermal conditions, a geraniol-derived substrate underwent the rearrangement with good stereoselectivity. The absolute configuration of the newly formed all-carbon quaternary stereocenter was confirmed by the total synthesis of (+)-bakuchiol from the rearrangement product.