MilliporeSigma
  • Comparison of capillary zone electrophoresis and isotachophoresis determination of dimethindene enantiomers in pharmaceuticals using charged carboxyethyl-beta-cyclodextrin as a chiral selector.
Comparison of capillary zone electrophoresis and isotachophoresis determination of dimethindene enantiomers in pharmaceuticals using charged carboxyethyl-beta-cyclodextrin as a chiral selector.
Methods and findings in experimental and clinical pharmacology (2007-01-04)
P Mikus, P Kubacak, I Valaskova, E Havranek
ABSTRACT
Charged cyclodextrin-mediated capillary electrophoretic methods (capillary zone electrophoresis [CZE] and isotachophoresis [ITP]) in a hydrodynamically closed separation system with suppressed electroosmotic flow and UV absorbance photometric detection (for CZE) or conductivity detection (for ITP) were developed for a highly effective separation and quantitation of dimethindene enantiomers in various pharmaceutical formulations (solution, gel, capsules). Optimized electrolyte systems were based on low-mobility buffer constituents (epsilon-aminocaproic acid, beta-alanine, potassium acetate, acetic acid), negatively charged (at pH > 4.5) carboxyethyl-beta-cyclodextrin (CE-beta-CD), serving as chiral selector, and methylhydroxyethylcellulose, serving as an electroosmotic flow suppressor. Complete enantioseparations of dimethindene in the presence of low concentrations of CE-beta-CD (2.5 mg/ml in CZE, 6.0 mg/ml in ITP) clearly indicated a role of charge of this chiral selector in enantioresolution. It also outlined the potential of charged CD-derivatives as chiral selectors for various CE techniques. The proposed methods were successfully validated, appraising parameters of sensitivity, linearity, precision, accuracy/recovery, and robustness, and then, they were applied to pharmaceutical samples. Consistent results obtained by both CE methods indicate their usefulness for routine use.
Product Description
Dimetindene maleate, European Pharmacopoeia (EP) Reference Standard