MilliporeSigma
  • [Study on condition for extraction of arctiin from fruits of Arctium lappa using supercritical fluid extraction].
[Study on condition for extraction of arctiin from fruits of Arctium lappa using supercritical fluid extraction].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-10-20)
Wen-hong Dong, Ben Liu
ABSTRACT
To study the feasibility of supercritical fluid extraction (SFE) for arctiin from the fruits of Arctium lappa. The extracts were analyzed by HPLC, optimum extraction conditions were studied by orthogonal tests. The optimal extraction conditions were: pressure 40 MPa, temperature 70 degrees C, using methanol as modifier carrier at the rate of 0.55 mL x min(-1), static extraction time 5 min, dynamic extraction 30 min, flow rate of CO2 2 L x min(-1). SFE has the superiority of adjustable polarity, and has the ability of extracting arctiin.
Sigma-Aldrich
Arctiin, ≥95% (LC/MS-ELSD)