MilliporeSigma
  • DNA-binding affinities and sequence selectivity of quaternary benzophenanthridine alkaloids sanguinarine, chelerythrine, and nitidine.
DNA-binding affinities and sequence selectivity of quaternary benzophenanthridine alkaloids sanguinarine, chelerythrine, and nitidine.
Bioorganic & medicinal chemistry (2006-05-30)
Li-Ping Bai, Zhong-Zhen Zhao, Zongwei Cai, Zhi-Hong Jiang
ABSTRACT
A comparative study on the intercalating binding of sanguinarine, chelerythrine, and nitidine with CT DNA, poly(dG-dC).poly(dG-dC), poly(dA-dT).poly(dA-dT), and seven sequence-designed double-stranded oligodeoxynucleotides has been performed using fluorometric and spectrophotometric techniques, aiming at providing insights into their sequence selectivity for DNA-binding. The results show that both sanguinarine and nitidine bind preferentially to DNA containing alternating GC base pairs [d(TGCGCA)(2)], while chelerythrine exhibits quite distinct sequence selectivity from sanguinarine, which shows a high specificity for DNA containing contiguous GC base pairs [5'-TGGGGA-3'/3'-ACCCCT-5'].