MilliporeSigma
  • Thiocolchicine-podophyllotoxin conjugates: dynamic libraries based on disulfide exchange reaction.
Thiocolchicine-podophyllotoxin conjugates: dynamic libraries based on disulfide exchange reaction.
The Journal of organic chemistry (2006-03-25)
Bruno Danieli, Alessandra Giardini, Giordano Lesma, Daniele Passarella, Bruno Peretto, Alessandro Sacchetti, Alessandra Silvani, Graziella Pratesi, Franco Zunino
ABSTRACT
A dynamic combinatorial library of thiocolchicine-podophyllotoxin derivatives based on the disulfide bond exchange reaction is described. The influence of a biological target on the composition of the reaction mixture has been demonstrated. Use of high-resolution ESI mass spectrometry to evaluate the composition of the mixture shows promise for the design of new large libraries. The biological evaluation demonstrates that formation of a divalent compound affords a new chemical entity whose biological activity is not merely the sum of the single ligands activities, thus reflecting a different interaction with the biological target.
Sigma-Aldrich
Thiocolchicine, ≥97% (HPLC)