MilliporeSigma
  • Effects of tetraiodothyronine and triiodothyronine on hamster cheek pouch microcirculation.
Effects of tetraiodothyronine and triiodothyronine on hamster cheek pouch microcirculation.
American journal of physiology. Heart and circulatory physiology (2004-12-04)
A Colantuoni, P L Marchiafava, D Lapi, F S Forini, G Iervasi
ABSTRACT
The aim of the present study was to assess the effects of topically applied triiodothyronine (T(3)) and thyroxine (T(4)) on the arterioles of hamster cheek pouch microcirculation in vivo. Microvessels were visualized using a fluorescent microscopy technique. Topical application of T(3) (3.08, 30.8, 61.5, 307, 615, and 6,150 nM/l) consistently induced dose-dependent dilation of arterioles within 2.0 +/- 0.5 min of administration. The application of T(4) (150, 257, 514, and 5,140 nM/l) caused different dose-dependent effects: dilation at the three lower doses within 16 +/- 2 min and rhythmic diameter changes at the highest dose. Aging of hamsters did not alter the arteriolar responses to T(3) and T(4). T(3)-induced dilation was countered by the inhibition of nitric oxide synthase with N(G)-nitro-L-arginine-methyl ester or N(G)-nitro-L-arginine. Iopanoic acid (IPA), which inhibits types I and II 5'-deiodinase, abolished the dilation elicited by 514 nM T(4) but did not affect T(3)-dependent dilation. 6-Propyl-2-thiouracil (PTU), which inhibits type I 5'-deiodinase only, did not affect the dilation induced by T(4). IPA and PTU did not impair arteriolar dilation induced by acetylcholine or sodium nitroprusside. These results indicate that T(3) induces arteriolar dilation, likely through nitric oxide release. The local conversion of T(4) to T(3) appears to be crucial for the dilation induced by T(4).
Product Description
Iopanoic acid, European Pharmacopoeia (EP) Reference Standard