MilliporeSigma
  • Inhibition of Naja nigricolis venom acidic phospholipase A2 catalysed hydrolysis of ghost red blood cells by columbin.
Inhibition of Naja nigricolis venom acidic phospholipase A2 catalysed hydrolysis of ghost red blood cells by columbin.
Journal of enzyme inhibition and medicinal chemistry (2002-10-09)
A J Nok, E Balogun, J A Lori, M S Abubakar
ABSTRACT
The inhibitory effects of a naturally occurring diterpenoid furanolactone, columbin, on partially purified acidic phospholipase A2 (PLA2) from Naja nigricolis was investigated. Columbin inhibited the N. nigricolis PLA2 in a dose related pattern with an IC50 value of 2.5 microM. Double reciprocal plots of initial velocity data of inhibition by columbin revealed a non-competitive pattern. The KM remained constant at 19 microM, while the Vmax changed from 54 micromoles/min/mg to 32 micromoles/min/mg and 20 micromoles/min/mg in the presence of 2 and 10 microM of columbin, respectively. Extrapolated Ki values were 3 and 6.28 microM at 2 and 10 microM inhibitor, respectively. Columbin also inhibited PLA2 hydrolysis of ghost RBC in a dose-dependent fashion. At least 70% suppression of PLA2-catalysed haemolysis of RBC was observed in the presence of 2 microM columbin.
Sigma-Aldrich
Columbin, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.