MilliporeSigma
  • Quantitative determination of liriodenine and moupinamide in five species of Mollinedia by high performance liquid chromatography.
Quantitative determination of liriodenine and moupinamide in five species of Mollinedia by high performance liquid chromatography.
Phytochemical analysis : PCA (2001-11-16)
D L Moreira, G G Leitão
ABSTRACT
A simple method for the resolution and subsequent quantitative determination of liriodenine and moupinamide in Mollinedia species was developed using reversed-phase HPLC based on an octadecyl silane-packed column eluted with gradients of methanol and water.
Sigma-Aldrich
Moupinamide, ≥95% (LC/MS-ELSD)