MilliporeSigma
  • Estrogen-related receptor gamma modulates energy metabolism target genes in human trophoblast.
Estrogen-related receptor gamma modulates energy metabolism target genes in human trophoblast.
Placenta (2012-07-06)
D Poidatz, E Dos Santos, A Brulé, P De Mazancourt, M N Dieudonné
ABSTRACT
Placenta growth and functions depend on correct trophoblast migration, proliferation, and differentiation. The placenta has a critical role in gas and nutrient transport. To accomplish these numerous functions, the placenta depends on a highly efficient energy metabolism control. Recent studies showed that the orphan nuclear receptor Estrogen-Related Receptor gamma (ERRγ) is highly expressed in human placentas. As ERRγ has been described as a major energy metabolism regulator, we investigated ERRγ expression and putative roles on energy homeostasis in human trophoblast from first trimester placentas. First, we showed that ERRγ expression level increased during pregnancy and that ERRγ was more abundant in villous than in extravillous trophoblasts. We also observed that ERRγ expression increased during trophoblast differentiation. Second, we demonstrated that mitochondrial biogenesis and expression of some energy metabolism target genes decreased when ERRγ expression was impaired. Altogether, these results suggest that ERRγ could be implicated in the energy metabolism regulation of human trophoblasts.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.