MilliporeSigma
  • Structure-activity relationships of bisphenol A analogs at estrogen receptors (ERs): discovery of an ERα-selective antagonist.
Structure-activity relationships of bisphenol A analogs at estrogen receptors (ERs): discovery of an ERα-selective antagonist.
Bioorganic & medicinal chemistry letters (2013-06-19)
Keisuke Maruyama, Masaharu Nakamura, Shusuke Tomoshige, Kazuyuki Sugita, Makoto Makishima, Yuichi Hashimoto, Minoru Ishikawa
ABSTRACT
Our multi-template approach for drug discovery, focusing on protein targets with similar fold structures, has yielded lead compounds for various targets. We have also shown that a diphenylmethane skeleton can serve as a surrogate for a steroid skeleton. Here, on the basis of those ideas, we hypothesized that the diphenylmethane derivative bisphenol A (BPA) would bind to the ligand-binding domain of estrogen receptors (ERs) in a similar manner to estradiol and act as a steroid surrogate. To test this idea, we synthesized a series of BPA analogs and evaluated their structure-activity relationships, focusing on agonistic/antagonistic activities at ERs and ERα/ERβ subtype selectivity. Among the compounds examined, 18 was found to be a potent ERα-antagonist with high selectivity over ERβ and androgen receptor under our assay conditions. A computational docking study suggested that 18 would bind to the antagonistic conformation of ERα. ERα-selective antagonists, such as 18, are candidate agents for treatment of breast cancer.