MilliporeSigma
  • Photoremovable protecting groups as controlled-release device for sex pheromone.
Photoremovable protecting groups as controlled-release device for sex pheromone.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2012-10-31)
Sanghamitra Atta, Mohammed Ikbal, Nishitha Boda, Samiran S Gauri, N D Pradeep Singh
ABSTRACT
Photoremovable protecting groups (PRPGs) were demonstrated as a delivery device for controlled release of pheromone under both UV light (≥350 nm) and direct sunlight irradiation. In the present work, (Z)-11-hexadecen-1-ol (sex pheromone of Chilo infuscatellus Snellen) was chemically caged by four different photoremovable protecting groups (7-hydroxy-4-hydroxymethylcoumarin, 1-pyrenemethanol, 9-anthracenemethanol and 2-(hydroxymethyl)anthraquinone) individually. Photophysical studies showed that the caged pheromone with coumarin, pyrene and anthracene derivatives exhibited strong fluorescence. Controlled release of (Z)-11-hexadecen-1-ol was achieved by irradiating the caged compounds in aqueous ethanol both under UV and sunlight. Further, to mimic the environmental conditions, controlled release of (Z)-11-hexadecen-1-ol was also studied in soil medium under direct sunlight. Thermogravimetric analysis showed that caging of (Z)-11-hexadecen-1-ol by PRPGs significantly reduced its volatility. Bioassay experiments indicated that PRPGs are harmless to soil bacteria (Azotobacter sp. and Pseudomonas aeruginosa) and in vitro cytotoxicity studies on eukaryotic L929 cells showed that PRPGs are also non-toxic. Field bioassays were performed using caged pheromone against maize stalk borer (Chilo partellus) and the results showed that the caged alcohol is effective in a number of moths catches instead of free alcohol in a blend for a longer period of time. Our studies indicated that use of PRPGs as delivery device for controlled release of pheromone by sunlight holds great interest for field applications.
Sigma-Aldrich
2-(Hydroxymethyl)anthraquinone, 97%
Pricing and availability is not currently available.