MilliporeSigma
  • Dissociative double ionization of 1-bromo-2-chloroethane irradiated by an intense femtosecond laser field.
Dissociative double ionization of 1-bromo-2-chloroethane irradiated by an intense femtosecond laser field.
The Journal of chemical physics (2011-08-17)
Yan Yang, Lulu Fan, Shengzhi Sun, Jian Zhang, Yuting Chen, Shian Zhang, Tianqing Jia, Zhenrong Sun
ABSTRACT
The dissociative double ionization and multi-photon ionization of 1-bromo-2-chloroethane (BCE) irradiated by the 800 nm femtosecond laser field have been investigated by dc-slice imaging technology. The charged parent ion ratio [BCE(2+)]/[BCE(+)] was measured, and the corresponding ionization process including non-sequential double ionization and sequential double ionization was analyzed. The sliced images of different photo-dissociated ions were detected, and the corresponding kinetic energy release (KER) distributions were calculated and extracted. Furthermore, the dissociative double ionization channels, attributed to the cleavage of the C-C, C-Br, and C-Cl bonds by the Coulombic repulsive forces, were discussed, and the revised equilibrium distance R(e)*, the energy ratio E(exp)/E(coul), and the value a=√(R(e)*)/(E(exp)/E(coul)) were calculated.
Sigma-Aldrich
1-Bromo-2-chloroethane, 98%