MilliporeSigma
  • A unique mRNA species for a regulatory subunit of cAMP-dependent protein kinase is specifically induced in haploid germ cells.
A unique mRNA species for a regulatory subunit of cAMP-dependent protein kinase is specifically induced in haploid germ cells.
FEBS letters (1988-03-14)
O Oyen, J D Scott, G G Cadd, G S McKnight, E G Krebs, V Hansson, T Jahnsen
ABSTRACT
Cyclic AMP (cAMP) and its action by way of cAMP-dependent protein kinase is important for sperm motility. Previous studies on germ cells have demonstrated a selective decrease in the amount of type I cAMP-dependent protein kinase during spermatid development, and that type II was the major form present in elongating spermatids and in mature sperm. This would indicate activation of a gene in haploid germ cells, encoding a regulatory subunit of type II protein kinase. However, haploid expression of such a gene has so far not been shown. In the present study we demonstrate high-levelled expression of a unique mRNA species for a specific regulatory subunit of type II cAMP-dependent protein kinase at late stages of spermatogenesis, i.e. during spermatid elongation.
Sigma-Aldrich
Lactoferrin from bovine milk, ≥85% (SDS-PAGE)