MilliporeSigma
  • Synthesis of trans-1,3-diaryl-2-(5-methylisoxazol-3-yl)-2,3-dihydro-1H-naphtho[1,2-e][1,3]oxazines via bismuth(III)-catalyzed one-pot pseudo-four component reaction.
Synthesis of trans-1,3-diaryl-2-(5-methylisoxazol-3-yl)-2,3-dihydro-1H-naphtho[1,2-e][1,3]oxazines via bismuth(III)-catalyzed one-pot pseudo-four component reaction.
Molecular diversity (2012-10-24)
Mehdi Shafiee, Ahmad Reza Khosropour, Iraj Mohammadpoor-Baltork, Majid Moghadam, Shahram Tangestaninejad, Valiollah Mirkhani, Hamid Reza Khavasi
ABSTRACT
An expeditious, straightforward and efficient synthesis of diversely naphtho[1,2-e][1,3]oxazines via one-pot condensation reaction of β- naphthol, 3-amino-5-methylisoxazole and arylaldehydes catalyzed by bismuth(III) trifluoromethanesulfonate is described. The reaction preferentially afforded 1,3-trans oxazines.
Sigma-Aldrich
3-Amino-5-methylisoxazole, ≥97%
Pricing and availability is not currently available.