MilliporeSigma
  • Evidence of superoxide radical contribution to demineralization of sulfamethoxazole by visible-light-driven Bi2O3/Bi2O2CO3/Sr6Bi2O9 photocatalyst.
Evidence of superoxide radical contribution to demineralization of sulfamethoxazole by visible-light-driven Bi2O3/Bi2O2CO3/Sr6Bi2O9 photocatalyst.
Journal of hazardous materials (2013-10-22)
Shiyuan Ding, Junfeng Niu, Yueping Bao, Lijuan Hu
ABSTRACT
Photocatalytic degradation of sulfamethoxazole (SMX) was investigated using Bi2O3/Bi2O2CO3/Sr6Bi2O9 (BSO) photocatalyst under visible light (>420 nm) irradiation. The photochemical degradation of SMX followed pseudo-first-order kinetics. The reaction kinetics was determined as a function of initial SMX concentrations (5-20 mg L(-1)), initial pH (3-11) and BSO concentrations (6-600 mg L(-1)). Approximately, 90% of SMX (10 mg L(-1)) degradation and 36% of TOC reduction were achieved at pH 7.0 after 120 min irradiation. The main mineralization products, including NH4(+), NO3(-), SO4(2-) and CO2, as well as intermediates 3-amino-5-methylisoxazole (AMI), p-benzoquinone (BZQ), and sulfanilic acid (SNA) were detected in aqueous solution. The formation of O2(*-) radical was evidenced by using electron spin resonance and a chemiluminescent probe, luminal. A possible degradation mechanism involving excitation of BSO, followed by charge injection into the BSO conduction band and formation of reactive superoxide radical (O2(*-)) was proposed for the mineralization of SMX. During the reaction, the O2(*-) radical attacks the sulfone moiety and causes the cleavage of the SN bond, which leads to the formation of two sub-structure analogs, AMI and SNA.
Sigma-Aldrich
3-Amino-5-methylisoxazole, ≥97%
Pricing and availability is not currently available.