MilliporeSigma
  • Synthesis of a paraffin phase change material microencapsulated in a siloxane polymer.
Synthesis of a paraffin phase change material microencapsulated in a siloxane polymer.
Colloid and polymer science (2013-03-06)
Witold Fortuniak, Stanislaw Slomkowski, Julian Chojnowski, Jan Kurjata, Adam Tracz, Urszula Mizerska
ABSTRACT
The coemulsification method suitable for the formulation of microcapsules of
Sigma-Aldrich
Eicosane, 99%