MilliporeSigma
  • The scalable pinacol coupling reaction utilizing the inorganic electride [Ca2N](+)·e(-) as an electron donor.
The scalable pinacol coupling reaction utilizing the inorganic electride [Ca2N](+)·e(-) as an electron donor.
Chemical communications (Cambridge, England) (2014-04-02)
Ye Ji Kim, Sun Min Kim, Hideo Hosono, Jung Woon Yang, Sung Wng Kim
ABSTRACT
The scalable pinacol coupling reaction is realized utilizing the inorganic electride [Ca2N](+)·e(-) as an electron donor in organic solvents. The bond cleavages of the [Ca2N](+) layers by methanol play a vital role in transferring anionic electrons to electrophilic aldehydes, accompanying the formation of Ca(OMe)2 and ammonia.
Sigma-Aldrich
Pinacol, 98%
Pricing and availability is not currently available.