MilliporeSigma
  • Polysaccharide-derived chiral stationary phases in capillary electrochromatography enantioseparations.
Polysaccharide-derived chiral stationary phases in capillary electrochromatography enantioseparations.
Methods in molecular biology (Clifton, N.J.) (2013-01-04)
Zhenbin Zhang, Hanfa Zou, Junjie Ou
ABSTRACT
Capillary electrochromatography (CEC) has been shown to be a promising technique for miniaturized enantioseparations. Here we present the preparation and application of positively charged cellulose derivative-based chiral stationary phases (CSPs) for the CEC separation of enantiomers. The cellulose derivatives were chemically immobilized onto diethylenetriaminopropylated silica gel using tolylene-2,4-diisocyanate as a spacer. The separation of enantiomers in CEC can be performed on the positively charged CSPs with nonaqueous and aqueous mobile phases.
Sigma-Aldrich
Tolylene-2,4-diisocyanate, technical grade, 80%
Pricing and availability is not currently available.