MilliporeSigma
  • Carbonic anhydrase inhibitors: inhibition of isozymes I, II and IV by sulfamide and sulfamic acid derivatives.
Carbonic anhydrase inhibitors: inhibition of isozymes I, II and IV by sulfamide and sulfamic acid derivatives.
Journal of enzyme inhibition (2000-10-13)
A Scozzafava, M D Banciu, A Popescu, C T Supuran
ABSTRACT
Sulfamide and sulfamic acid are the simplest compounds containing the SO2NH2 moiety, responsible for binding to the Zn(II) ion within carbonic anhydrase (CA, EC 4.2.1.1) active site, and thus acting as inhibitors of the many CA isozymes presently known. Here we describe two novel classes of CA inhibitors obtained by derivatizations of the lead molecules mentioned above. The new compounds, possessing the general formula RSO2NH-SO2X (X = OH, NH2), were obtained by reaction of sulfamide or sulfamic acid with alkyl/arylsulfonyl halides or arylsulfonyl isocyanates. A smaller series of derivatives has been obtained by reaction of aromatic aldehydes with sulfamide, leading to Schiff bases of the type ArCH = NSO2NH2. All the new compounds act as strong inhibitors of isozymes I, II and IV of carbonic anhydrase. Their mechanism of CA inhibition is also discussed based on electronic spectroscopic measurements on adducts with the Co(II)-substituted enzyme. These experiments led to the conclusion that the new inhibitors are directly coordinated (in a monodentate manner) to the metal ion within the enzyme active site, similarly to the classical inhibitors, the aromatic/heterocyclic sulfonamides.
Sigma-Aldrich
Sulfamide, 99%