MilliporeSigma
  • Design, synthesis, and biological characterization of a caspase 3/7 selective isatin labeled with 2-[18F]fluoroethylazide.
Design, synthesis, and biological characterization of a caspase 3/7 selective isatin labeled with 2-[18F]fluoroethylazide.
Journal of medicinal chemistry (2008-12-04)
Graham Smith, Matthias Glaser, Meg Perumal, Quang-De Nguyen, Bo Shan, Erik Arstad, Eric O Aboagye
ABSTRACT
Imaging of programmed cell death (apoptosis) is important in the assessment of therapeutic response in oncology and for diagnosis in cardiac and neurodegenerative disorders. The executioner caspases 3 and 7 ultimately effect cellular death, thus providing selective molecular targets for in vivo quantification of apoptosis. To realize this potential, we aimed to develop 18F-labeled isatin sulfonamides with high metabolic stability and moderate lipophilicity while retaining selectivity and affinity for caspase 3/7. A small library of isatins modified with fluorinated aromatic groups and heterocycles was synthesized. A lead compound incorporating 2'-fluoroethyl-1,2,3-triazole was identified with subnanomolar affinity for caspase 3. "Click labeling" provided the 18F-labeled tracer in 65 +/- 6% decay-corrected radiochemical yield from 2-[18F]fluoroethylazide. The compound showed high stability in vivo with rapid uptake and elimination in healthy tissues and tumor. The novel 18F-labeled isatin is a candidate radiotracer for further preclinical evaluation for imaging of apoptosis.
Sigma-Aldrich
4-Fluorobenzyl bromide, 97%
Pricing and availability is not currently available.