MilliporeSigma
  • Alternating hydrophilic and hydrophobic pockets in the channel structures of organostannoxane prismanes: preferential confinement of guest molecules.
Alternating hydrophilic and hydrophobic pockets in the channel structures of organostannoxane prismanes: preferential confinement of guest molecules.
Journal of the American Chemical Society (2005-08-18)
Vadapalli Chandrasekhar, Pakkirisamy Thilagar, James F Bickley, Alexander Steiner
ABSTRACT
A hydroxyl-rich hexameric organooxotin prismane has been prepared by reaction of n-BuSn(O)OH with 9-hydroxy-9-fluorenecarboxylic acid. The supramolecular structure of this cage shows channels with hydrophobic and hydrophilic segments, which selectively entrap guest molecules.
Sigma-Aldrich
9-Hydroxy-9-fluorenecarboxylic acid, 96%
Pricing and availability is not currently available.