MilliporeSigma
  • Downregulation of microRNA-30 facilitates podocyte injury and is prevented by glucocorticoids.
Downregulation of microRNA-30 facilitates podocyte injury and is prevented by glucocorticoids.
Journal of the American Society of Nephrology : JASN (2013-09-14)
Junnan Wu, Chunxia Zheng, Yun Fan, Caihong Zeng, Zhaohong Chen, Weisong Qin, Changming Zhang, Wanfen Zhang, Xiao Wang, Xiaodong Zhu, Mingchao Zhang, Ke Zen, Zhihong Liu
ABSTRACT
MicroRNAs (miRNAs) are essential for podocyte homeostasis, and the miR-30 family may be responsible for this action. However, the exact roles and clinical relevance of miR-30s remain unknown. In this study, we examined the expression of the miR-30 family in the podocytes of patients with FSGS and found that all members are downregulated. Treating cultured human podocytes with TGF-β, LPS, or puromycin aminonucleoside (PAN) also downregulated the miR-30 family. Podocyte cytoskeletal damage and apoptosis caused by treatment with TGF-β or PAN were ameliorated by exogenous miR-30 expression and aggravated by miR-30 knockdown. Moreover, we found that miR-30s exert their protective roles by direct inhibition of Notch1 and p53, which mediate podocyte injury. In rats, treatment with PAN substantially downregulated podocyte miR-30s and induced proteinuria and podocyte injury; however, transfer of exogenous miR-30a to podocytes of PAN-treated rats ameliorated proteinuria and podocyte injury and reduced Notch1 activation. Finally, we demonstrated that glucocorticoid treatment maintains miR-30 expression in cultured podocytes treated with TGF-β, LPS, or PAN and in the podocytes of PAN-treated rats. Glucocorticoid-sustained miR-30 expression associated with reduced Notch1 activation and alleviated podocyte damage. Taken together, these findings demonstrate that miR-30s protect podocytes by targeting Notch1 and p53 and that the loss of miR-30s facilitates podocyte injury. In addition, sustained miR-30 expression may be a novel mechanism underlying the therapeutic effectiveness of glucocorticoids in treating podocytopathy.
Sigma-Aldrich
Puromycin aminonucleoside
Pricing and availability is not currently available.