MilliporeSigma
  • Detectable urogenital schistosome DNA and cervical abnormalities 6 months after single-dose praziquantel in women with Schistosoma haematobium infection.
Detectable urogenital schistosome DNA and cervical abnormalities 6 months after single-dose praziquantel in women with Schistosoma haematobium infection.
Tropical medicine & international health : TM & IH (2013-08-14)
Jennifer A Downs, Rodrick Kabangila, Jaco J Verweij, Hyasinta Jaka, Robert N Peck, Samuel E Kalluvya, John M Changalucha, Warren D Johnson, Lisette van Lieshout, Daniel W Fitzgerald
ABSTRACT
We explored response to single-dose praziquantel therapy in a cohort of 33 women with Schistosoma haematobium infection in rural Mwanza, Tanzania. Women with S. haematobium infection confirmed both by eggs in urine and by polymerase chain reaction (PCR) received single-dose praziquantel and treatment of concomitant sexually transmitted infections. Macroscopic cervical abnormalities were also quantified. After 6 months, microscopically detectable egg excretion was eliminated, but 8 of 33 women (24%) were persistently positive for S. haematobium by PCR, and 11 (33%) had cervical abnormalities potentially attributable to schistosomiasis. This suggests that praziquantel treatment more frequently than every 6 months may be necessary for complete elimination of the parasite and prevention of genital tissue pathology. This aggressive therapy may in turn play a key role decreasing HIV susceptibility in millions of people living in regions in which S. haematobium is endemic.
Product Description
Supelco
Praziquantel, VETRANAL®, analytical standard