MilliporeSigma
  • A segment of the 5' nontranslated region of encephalomyocarditis virus RNA directs internal entry of ribosomes during in vitro translation.
A segment of the 5' nontranslated region of encephalomyocarditis virus RNA directs internal entry of ribosomes during in vitro translation.
Journal of virology (1988-08-01)
S K Jang, H G Kräusslich, M J Nicklin, G M Duke, A C Palmenberg, E Wimmer
ABSTRACT
Picornavirus RNAs are uncapped messengers and have unusually long 5' nontranslated regions (5'NTRs) which contain many noninitiating AUG triplets. The translational efficiency of different picornavirus RNAs varies between different cell-free extracts and even in the same extract, such as micrococcal nuclease-treated rabbit reticulocyte lysates. The effect of the poliovirus 5'NTR on in vitro translation was compared with that of the 5'NTR of encephalomyocarditis virus by the use of synthetic mRNAs, micrococcal nuclease-treated HeLa cell extracts, and rabbit reticulocyte lysates. Artificial mono- and dicistronic mRNAs synthesized with T7 RNA polymerase were used to investigate whether the 5'NTR of encephalomyocarditis virus RNA contains a potential internal ribosomal entry site. The sequence between nucleotides 260 and 484 in the 5'NTR of encephalomyocarditis RNA was found to play a critical role in the efficient translation in both mono- and dicistronic mRNAs. Our data suggest that an internal ribosomal entry site resides in this region.
Product Description
Millipore
EZview Red Glutathione Affinity Gel
Pricing and availability is not currently available.
Sigma-Aldrich
pBICEP-CMV-3 Expression Vector
Pricing and availability is not currently available.