MilliporeSigma
  • Casein aggregates built step-by-step on charged polyelectrolyte film surfaces are calcium phosphate-cemented.
Casein aggregates built step-by-step on charged polyelectrolyte film surfaces are calcium phosphate-cemented.
The Journal of biological chemistry (2010-10-06)
Krisztina Nagy, Ana-Maria Pilbat, Géza Groma, Balázs Szalontai, Frédéric J G Cuisinier
ABSTRACT
The possible mechanism of casein aggregation and micelle buildup was studied in a new approach by letting α-casein adsorb from low concentration (0.1 mg·ml(-1)) solutions onto the charged surfaces of polyelectrolyte films. It was found that α-casein could adsorb onto both positively and negatively charged surfaces. However, only when its negative phosphoseryl clusters remained free, i.e. when it adsorbed onto a negative surface, could calcium phosphate (CaP) nanoclusters bind to the casein molecules. Once the CaP clusters were in place, step-by-step building of multilayered casein architectures became possible. The presence of CaP was essential; neither Ca(2+) nor phosphate could alone facilitate casein aggregation. Thus, it seems that CaP is the organizing motive in the casein micelle formation. Atomic force microscopy revealed that even a single adsorbed casein layer was composed of very small (in the range of tens of nanometers) spherical forms. The stiffness of the adsorbed casein layer largely increased in the presence of CaP. On this basis, we can imagine that casein micelles emerge according to the following scheme. The amphipathic casein monomers aggregate into oligomers via hydrophobic interactions even in the absence of CaP. Full scale, CaP-carrying micelles could materialize by interlocking these casein oligomers with CaP nanoclusters. Such a mechanism would not contradict former experimental results and could offer a synthesis between the submicelle and the block copolymer models of casein micelles.
Sigma-Aldrich
Casein, Hammarsten bovine
Pricing and availability is not currently available.
Sigma-Aldrich
Casein, Hammarsten bovine, Suitable for manufacturing of diagnostic kits and reagents
Pricing and availability is not currently available.