MilliporeSigma
  • New 5-HT1A receptor agonists possessing 1,4-benzoxazepine scaffold exhibit highly potent anti-ischemic effects.
New 5-HT1A receptor agonists possessing 1,4-benzoxazepine scaffold exhibit highly potent anti-ischemic effects.
Bioorganic & medicinal chemistry letters (2001-03-07)
K Kamei, N Maeda, R Ogino, M Koyama, M Nakajima, T Tatsuoka, T Ohno, T Inoue
ABSTRACT
A series of new 3-substituted-4-(4-aminobutyl)-1,4-benzoxazepin-5(4H)-one derivatives (1-5) which showed a very high affinity for 5-HT1A receptor with good selectivity over dopamine D2 receptor was synthesized. Among these compounds, 3-chloro-4-[4-[4-(2-pyridinyl)-1,2,3,6-tetrahydropyridin-1-yl]butyl]-1,4-benzoxazepin-5(4H)-one (5: SUN N4057) exhibited remarkable neuroprotective activity in a transient middle cerebral artery occlusion (t-MCAO) model.
Sigma-Aldrich
2-Bromomethyl-1,3-dioxolane, 96%