MilliporeSigma
  • Fluorescence detection of surface-bound intermediates produced from UV photoreactivity of alkylsiloxane SAMs.
Fluorescence detection of surface-bound intermediates produced from UV photoreactivity of alkylsiloxane SAMs.
Journal of the American Chemical Society (2004-02-26)
Eric A McArthur, Tao Ye, Jason P Cross, Stéphane Petoud, Eric Borguet
ABSTRACT
Detection and quantification of submonolayer coverage surface species is not trivial. We have developed a novel method sensitive to surface-bound chemical functional groups as low as 10(11) molecules/cm(2) by specific covalent attachment of fluorescent chromophores. This enables the intermediates of the UV photochemistry of alkylsiloxane self-assembled monolayers to be identified.