MilliporeSigma
  • A new and efficient strategy for the synthesis of shimofuridin analogs: 2'-O-(4-O-stearoyl-alpha-L-fucopyranosyl)thymidine and -uridine.
A new and efficient strategy for the synthesis of shimofuridin analogs: 2'-O-(4-O-stearoyl-alpha-L-fucopyranosyl)thymidine and -uridine.
Carbohydrate research (2002-12-31)
Jun Ning, Ying Xing, Fanzuo Kong
ABSTRACT
Two shimofuridin analogs: 2'-O-(4-O-stearoyl-alpha-L-fucopyranosyl)thymidine (2) and -uridine (3) have been synthesized using D-arabinose, L-fucose, thymine, uracil, and stearoyl chloride as the starting materials. The synthetic procedures involve the facile preparation of 1-(3,5-di-O-benzyl-beta-D-ribofuranosyl)thymine (9) and -uracil (10) by coupling of 1,2-anhydro-3,5-di-O-benzyl-alpha-D-ribofuranose (8) with silylated thymine and uracil, and then stereoselective formation of the 1,2-cis (alpha) interglycoside bonds through condensation of the nucleoside derivatives 9 and 10 with 2-(2,3-di-O-benzyl-4-O-stearoyl-beta-L-fucopyranosylsulfonyl) pyrimidine (18). The 1,2-anhydro-3,5-di-O-benzyl-alpha-D-ribofuranose (8) was prepared by an improved procedure from D-arabinose.
Sigma-Aldrich
Stearoyl chloride, 97%