MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis and pharmacological evaluation of some 3-substituted octahydropyrido(2,1-c)(1,4)oxazines.
Synthesis and pharmacological evaluation of some 3-substituted octahydropyrido(2,1-c)(1,4)oxazines.
Journal of medicinal chemistry (1976-02-01)
T N Riley, G O Rankin
ABSTRACT
3-Phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers have been synthesized from (+/-)-, (+)-, and (-)-2-piperidinemethanol. Treatment of 2-piperidinemethanol with alpha-bromoacetophenone gave 3-hydroxy-3-phenyloctahydropyrido[2,1-c][1,4]oxazine which was readily converted to the 3-phenyl derivative by catalytic hydrogenolysis. These compounds were shown to possess a depressant action on the CNS which was quantitated in terms of reduction of locomotor activity in mice. Qualitative differences were noted in the central effects of the 3-phenyl compound and its hemiketal derivative. Further, qualitative differences in the effects of the diastereomers of the 3-phenyl compound on locomotor activity of mice were also noted. The results of this study suggest that the octahydropyrido[2,1-c]1,4]oxazine system may provide a useful molecular framework for the construction of agents exhibiting pharmacologically useful actions in the CNS.
Sigma-Aldrich
2-Piperidinemethanol, 97%