MilliporeSigma
  • Three novel 1D lanthanide-carboxylate polymeric complexes: syntheses, crystal structures and magnetic analyses.
Three novel 1D lanthanide-carboxylate polymeric complexes: syntheses, crystal structures and magnetic analyses.
Dalton transactions (Cambridge, England : 2003) (2013-05-01)
Jumei Tian, Bo Li, Xiaoying Zhang, Xiaolei Li, Xiaoliang Li, Jingping Zhang
ABSTRACT
Three novel one-dimensional (1D) lanthanide coordination complexes involving the 4-methyl-3-nitrobenzoic acid (HL) ligand, with the general formulae [Gd(L)3(H2O)(CH3OH)] (1), [Gd(L)3(H2O)2]·(4,4'-bpy)·CH3OH (2), [Dy(L)3(H2O)(CH3OH)] (3), have been synthesized by solvothermal reactions. These complexes have been structurally characterized by single-crystal X-ray diffraction, IR, PXRD, TGA, and elemental analysis. Complexes 1 and 3 are isostructural except for the distinction of the metal ion. Magnetic measurements indicate that complexes 1-3 show antiferromagnetic behaviors. Complex 3 behaves with slow relaxation of the magnetization, where the frequency-dependent out-of-phase signals are noticed. However, the characteristic maxima were not reached above 2 K under zero direct current (DC) fields. When a DC field of 5000 Oe was employed, the frequency dependent peaks of alternating current (AC) signals were obtained.